Learn More
Biomatik Corporation KIF20A Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50344
Encompass_Preferred
KIF20A Colorimetric Cell-Based ELISA Kit for detecting KIF20A in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O95235. Gene ID: 10112. Target Synonyms: Kinesin-like protein KIF20A; Rabkinesin-6; Rab6-interacting kinesin-like protein; GG10_2; RAB6KIFL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0615