Learn More
Biomatik Corporation Mammaglobin B Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50358
Encompass_Preferred
Mammaglobin B Colorimetric Cell-Based ELISA Kit for detecting Mammaglobin B in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75556. Gene ID: 4246. Target Synonyms: Mammaglobin-B; Mammaglobin-2; Lipophilin-C; Lacryglobin; Secretoglobin family 2A member 1; SCGB2A1; LIPHC; MGB2; UGB3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0628