Learn More
Biomatik Corporation MCM5 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50370
Encompass_Preferred
MCM5 Colorimetric Cell-Based ELISA Kit for detecting MCM5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P33992. Gene ID: 4174. Target Synonyms: CDC46; DNA replication licensing factor MCM5; minichromosome maintenance deficient protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0638