Learn More
Biomatik Corporation MMP-2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50396
Encompass_Preferred
MMP-2 Colorimetric Cell-Based ELISA Kit for detecting MMP-2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08253. Gene ID: 4313. Target Synonyms: 72 kDa gelatinase; 72 kDa type IV collagenase; CLG4; CLG4A; collagenase type IV-A; gelatinase A; matrix metalloproteinase-2; MMP-2; MMP-II; MONA; TBE-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0662