Learn More
Biomatik Corporation MMP-3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50398
Encompass_Preferred
MMP-3 Colorimetric Cell-Based ELISA Kit for detecting MMP-3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08254. Gene ID: 4314. Target Synonyms: Stromelysin-1; SL-1; Matrix metalloproteinase-3; MMP-3; Transin-1; STMY1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0664