Learn More
Biomatik Corporation Osteopontin Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50434
Encompass_Preferred
Osteopontin Colorimetric Cell-Based ELISA Kit for detecting Osteopontin in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10451. Gene ID: 6696. Target Synonyms: Bone sialoprotein 1; Nephropontin; OPN; OSTP; SPP-1; SPP1; Secreted phosphoprotein 1; Urinary stone protein; Uropontin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0689