Learn More
Biomatik Corporation PKCB Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50488
Encompass_Preferred
PKCB Colorimetric Cell-Based ELISA Kit for detecting PKCB in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05771. Gene ID: 5579. Target Synonyms: kinase PKC-beta; KPCB; PKC II; PKC-B; PKC-beta; PKC-II; PRKCB; PRKCB1; Protein kinase C beta type. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0740