Learn More
Biomatik Corporation Prostate Apoptosis Response protein-4 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50505
Encompass_Preferred
Prostate Apoptosis Response protein-4 Colorimetric Cell-Based ELISA Kit for detecting Prostate Apoptosis Response protein-4 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96IZ0. Gene ID: 5074. Target Synonyms: Par-4; PAR4; PRKC apoptosis WT1 regulator protein; PRKC; apoptosis; WT1; regulator; prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; prostate apoptosis response-4 protein; transcriptional repressor PAR4; WT1-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0753