Learn More
Biomatik Corporation RAD52 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50515
Encompass_Preferred
RAD52 Colorimetric Cell-Based ELISA Kit for detecting RAD52 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P43351. Gene ID: 5893. Target Synonyms: DNA repair protein RAD52 homolog; RA52. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0761