Learn More
Biomatik Corporation RCBTB1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50517
Encompass_Preferred
RCBTB1 Colorimetric Cell-Based ELISA Kit for detecting RCBTB1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8NDN9. Gene ID: 55213. Target Synonyms: RCC1 and BTB domain-containing protein 1; Regulator of chromosome condensation and BTB domain-containing protein 1; Chronic lymphocytic leukemia deletion region gene 7 protein; CLL deletion region gene 7 protein; CLLD7; E4.5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0763