Learn More
Biomatik Corporation Retinoic Acid Receptor β Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50522
Encompass_Preferred
Retinoic Acid Receptor β Colorimetric Cell-Based ELISA Kit for detecting Retinoic Acid Receptor β in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10826. Gene ID: 5915. Target Synonyms: HBV-activated protein; nuclear receptor 1B2; RAR-beta; RAR-epsilon; retinoic acid receptor beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0767