Learn More
Biomatik Corporation RFA2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50524
Encompass_Preferred
RFA2 Colorimetric Cell-Based ELISA Kit for detecting RFA2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15927. Gene ID: 6118. Target Synonyms: REPA2; replication factor-A protein 2; replication protein A 32 kDa subunit; replication protein A2; RF-A; RP-A; RPA2; RPA32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0769