Learn More
Biomatik Corporation Smad4 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50551
Encompass_Preferred
Smad4 Colorimetric Cell-Based ELISA Kit for detecting Smad4 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13485. Gene ID: 4089. Target Synonyms: Mothers against decapentaplegic homolog 4; MAD homolog 4; Mothers against DPP homolog 4; Deletion target in pancreatic carcinoma 4; SMAD family member 4; SMAD 4; Smad4; hSMAD4; SMAD4; DPC4; MADH4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0791