Learn More
Biomatik Corporation Stathmin 1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50568
Encompass_Preferred
Stathmin 1 Colorimetric Cell-Based ELISA Kit for detecting Stathmin 1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16949. Gene ID: 3925. Target Synonyms: LAG; LAP18; Leukemia-associated gene protein; Leukemia-associated phosphoprotein p18; Metablastin; Oncoprotein 18; Op18; Phosphoprotein p19; pp17; pp19; PR22; Pr22 protein; Prosolin; stathmin; STN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0805