Learn More
Biomatik Corporation TPH2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50598
Encompass_Preferred
TPH2 Colorimetric Cell-Based ELISA Kit for detecting TPH2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8IWU9. Gene ID: 121278. Target Synonyms: Neuronal tryptophan hydroxylase; NTPH; Tryptophan 5- monooxygenase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0832