Learn More
Biomatik Corporation WAVE1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50625
Encompass_Preferred
WAVE1 Colorimetric Cell-Based ELISA Kit for detecting WAVE1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92558. Gene ID: 8936. Target Synonyms: KIAA0269; SCAR1; Verprolin homology domain-containing protein 1; WAS1; WASF1; WASP-family protein member 1; Wiskott-Aldrich syndrome protein family member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0855