Learn More
Biomatik Corporation TBX10 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50674
Encompass_Preferred
TBX10 Colorimetric Cell-Based ELISA for detecting TBX10 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75333. Gene ID: 347853. Target Synonyms: T-box transcription factor TBX10; T-box protein 10; TBX10; TBX7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0900