Learn More
Biomatik Corporation GLU2B Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50706
Encompass_Preferred
GLU2B Colorimetric Cell-Based ELISA for detecting GLU2B in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P14314. Gene ID: 5589. Target Synonyms: 80K-H; AGE-R2; AGE-binding receptor 2; G19P1; GLU2B; PCLD; PKCSH; glucosidase II beta subunit; glucosidase II; beta subunit; hepatocystin; protein kinase C substrate; 60.1 kDa protein; heavy chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0932