Learn More
Biomatik Corporation AKAP5 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50724
Encompass_Preferred
AKAP5 Colorimetric Cell-Based ELISA for detecting AKAP5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P24588. Gene ID: 9495. Target Synonyms: A kinase (PRKA) anchor protein 5; A-kinase anchor 5; A-kinase anchor 79 kDa; AKAP 79; AKAP75; AKAP79; cAMP-dependent protein kinase regulatory subunit II high affinity-binding; H21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0950