Learn More
Biomatik Corporation NFYB Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50726
Encompass_Preferred
NFYB Colorimetric Cell-Based ELISA for detecting NFYB in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25208. Gene ID: 4801. Target Synonyms: Nuclear transcription factor Y subunit beta; Nuclear transcription factor Y subunit B; NF-YB; CAAT-box DNA-binding protein subunit B; HAP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0952