Learn More
Biomatik Corporation PPIF Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50734
Encompass_Preferred
PPIF Colorimetric Cell-Based ELISA for detecting PPIF in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P30405. Gene ID: 10105. Target Synonyms: Peptidyl-prolyl cis-trans isomerase; mitochondrial; PPIase; Rotamase; Cyclophilin F; PPIF; CYP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0959