Learn More
Biomatik Corporation HOXD12 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50740
Encompass_Preferred
HOXD12 Colorimetric Cell-Based ELISA for detecting HOXD12 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35452. Gene ID: 3238. Target Synonyms: Homeobox protein Hox-D12; Hox-4H; HOXD12; HOX4H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0965