Learn More
Biomatik Corporation GRK3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50743
Encompass_Preferred
GRK3 Colorimetric Cell-Based ELISA for detecting GRK3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35626. Gene ID: 157. Target Synonyms: ADRBK2; ARBK2; BARK2; G-protein coupled receptor kinase 3; beta-ARK-2; beta-adrenergic receptor kinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0968