Learn More
Biomatik Corporation ACVL1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50746
Encompass_Preferred
ACVL1 Colorimetric Cell-Based ELISA for detecting ACVL1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P37023. Gene ID: 94. Target Synonyms: ACVL1; ACVRL1; ACVRLK1; ALK-1; Activin receptor-like kinase 1; SKR3; TGF-B superfamily receptor type I; TSR-I; kinase ALK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0971