Learn More
Biomatik Corporation EFNA5 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50778
Encompass_Preferred
EFNA5 Colorimetric Cell-Based ELISA for detecting EFNA5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52803. Gene ID: 1946. Target Synonyms: EPH-related receptor tyrosine kinase ligand 7; LERK-7; AL-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1002