Learn More
Biomatik Corporation MEF2B Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50800
Encompass_Preferred
MEF2B Colorimetric Cell-Based ELISA for detecting MEF2B in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q02080. Gene ID: 4207. Target Synonyms: Myocyte-specific enhancer factor 2B; RSRFR2; Serum response factor-like protein 2 ; MEF2B ; XMEF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1023