Learn More
Biomatik Corporation AKAP13 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50817
Encompass_Preferred
AKAP13 Colorimetric Cell-Based ELISA for detecting AKAP13 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q12802. Gene ID: 11214. Target Synonyms: A-kinase anchor protein 13; AKAP-13; AKAP-Lbc; Breast cancer nuclear receptor-binding auxiliary protein; Guanine nucleotide exchange factor Lbc; Human thyroid-anchoring protein 31; Lymphoid blast crisis oncogene; LBC oncogene; Non-oncogenic Rho GTPase-specific GTP exchange factor; Protein kinase A-anchoring protein 13; p47 ; AKAP13 ; BRX; HT31; LBC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1039