Learn More
Biomatik Corporation DOK7 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50873
Encompass_Preferred
DOK7 Colorimetric Cell-Based ELISA for detecting DOK7 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q18PE1. Gene ID: 285489. Target Synonyms: Downstream of tyrosine kinase 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1092