Learn More
Biomatik Corporation DAK Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50875
Encompass_Preferred
DAK Colorimetric Cell-Based ELISA for detecting DAK in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q3LXA3. Gene ID: 26007. Target Synonyms: DHA kinase; Dihydroxyacetone kinase; Glycerone kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1094