Learn More
Biomatik Corporation ANKK1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50907
Encompass_Preferred
ANKK1 Colorimetric Cell-Based ELISA for detecting ANKK1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8NFD2. Gene ID: 255239. Target Synonyms: Protein kinase PKK2; X-kinase; Sugen kinase 288; SgK288. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1126