Learn More
Biomatik Corporation TF3C2 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50916
Encompass_Preferred
TF3C2 Colorimetric Cell-Based ELISA for detecting TF3C2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WUA4. Gene ID: 2976. Target Synonyms: Transcription factor IIIC subunit beta; TF3C-beta; TFIIIC 110 kDa subunit; TFIIIC110. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1135