Learn More
Biomatik Corporation MADD Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50918
Encompass_Preferred
MADD Colorimetric Cell-Based ELISA for detecting MADD in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WXG6. Gene ID: 8567. Target Synonyms: Differentially expressed in normal and neoplastic cells; Insulinoma glucagonoma clone 20; Rab3 GDP/GTP exchange factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1137