Learn More
Biomatik Corporation Cytochrome P450 2C8/9/18/19 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50943
Encompass_Preferred
Cytochrome P450 2C8/9/18/19 Colorimetric Cell-Based ELISA for detecting Cytochrome P450 2C8/9/18/19 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P33260; P10632; P11712; P33261. Gene ID: 1562; 1558; 1559; 1557. Target Synonyms: CP2CI; CPCI; CYPIIC18; P450-6B/29C; cytochrome P450 2C18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1161