Learn More
Biomatik Corporation Cytochrome P450 4B1 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50953
Encompass_Preferred
Cytochrome P450 4B1 Colorimetric Cell-Based ELISA for detecting Cytochrome P450 4B1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P13584. Gene ID: 1580. Target Synonyms: Cytochrome P450 4B1; CYPIVB1; P450-HP; CYP4B1; CP4B1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1171