Learn More
Biomatik Corporation CBX5 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50753
Encompass_Preferred
CBX5 Colorimetric Cell-Based ELISA for detecting CBX5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P45973. Gene ID: 23468. Target Synonyms: HP1 alpha; HP1A; antigen p25; chromobox protein homolog 5; heterochromatin protein 1 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0978