Learn More
Biomatik Corporation ID4 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50755
Encompass_Preferred
ID4 Colorimetric Cell-Based ELISA for detecting ID4 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P47928. Gene ID: 3400. Target Synonyms: DNA-binding protein inhibitor ID-4; Inhibitor of DNA binding 4; ID4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0980