Learn More
Biomatik Corporation Hexokinase-3 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50776
Encompass_Preferred
Hexokinase-3 Colorimetric Cell-Based ELISA for detecting Hexokinase-3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52790. Gene ID: 3101. Target Synonyms: Hexokinase type III; HK III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1000