Learn More
Biomatik Corporation Mammaglobin Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50359
Encompass_Preferred
Mammaglobin Colorimetric Cell-Based ELISA Kit for detecting Mammaglobin in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13296. Gene ID: 4250. Target Synonyms: Mammaglobin-A; Mammaglobin-1; Secretoglobin family 2A member 2; SCGB2A2; MGB1; UGB2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0629