Learn More
Biomatik Corporation mGluR4 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50380
Encompass_Preferred
mGluR4 Colorimetric Cell-Based ELISA Kit for detecting mGluR4 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14833. Gene ID: 2914. Target Synonyms: Metabotropic glutamate receptor 4; mGluR4; GPRC1D; MGLUR4; GRM4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0648