Learn More
Biomatik Corporation MKP-1/2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50388
Encompass_Preferred
MKP-1/2 Colorimetric Cell-Based ELISA Kit for detecting MKP-1/2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P28562; Q13115. Gene ID: 1843; 1846. Target Synonyms: 3CH134; CL100; Dual specificity protein phosphatase 1; Dual specificity protein phosphatase hVH1; DUS1; DUSP1; MAP kinase phosphatase-1; MKP1; Protein-tyrosine phosphatase 3CH134; Protein-tyrosine phosphatase CL100; Protein-tyrosine phosphatase ERP; PTPN10; PTPN16; VH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0655