Learn More
Biomatik Corporation MMP-8 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50400
Encompass_Preferred
MMP-8 Colorimetric Cell-Based ELISA Kit for detecting MMP-8 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22894. Gene ID: 4317. Target Synonyms: Neutrophil collagenase; Matrix metalloproteinase-8; MMP-8; PMNL collagenase; PMNL-CL; CLG1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0665