Learn More
Biomatik Corporation NM23 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50422
Encompass_Preferred
NM23 Colorimetric Cell-Based ELISA Kit for detecting NM23 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22392. Gene ID: 4831; 654364. Target Synonyms: c-myc purine-binding transcription factor PUF; NDK B; NDKB; NDP kinase B; NDPKB; NM23B; nucleoside diphosphate kinase B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0681