Learn More
Biomatik Corporation p47 phox Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50443
Encompass_Preferred
p47 phox Colorimetric Cell-Based ELISA Kit for detecting p47 phox in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P14598. Gene ID: 653361. Target Synonyms: 47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NCF-1; NCF-47K; NCF1; Neutrophil cytosol factor 1; Neutrophil cytosolic factor 1; Neutrophil NADPH oxidase factor 1; P47 phox; p47-phox. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0697