Learn More
Biomatik Corporation PDGFB Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50466
Encompass_Preferred
PDGFB Colorimetric Cell-Based ELISA Kit for detecting PDGFB in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01127. Gene ID: 5155. Target Synonyms: Platelet-derived growth factor subunit B; PDGF subunit B; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; PDGF-2; c-sis; PDGF2; SIS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0720