Learn More
Biomatik Corporation PGP9.5 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50474
Encompass_Preferred
PGP9.5 Colorimetric Cell-Based ELISA Kit for detecting PGP9.5 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09936. Gene ID: 7345. Target Synonyms: UCH-L1; neuron cytoplasmic protein 9.5; ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin thiolesterase L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0726