Learn More
Biomatik Corporation PKC ζ Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50486
Encompass_Preferred
PKC ζ Colorimetric Cell-Based ELISA Kit for detecting PKC ζ in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05513. Gene ID: 5590. Target Synonyms: KPCZ; PKC-zeta; PKC2; PRKCZ; Protein kinase C zeta type; kinase PKC-zeta; nPKC-zeta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0738