Learn More
Biomatik Corporation PKD1/PKC μ Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50490
Encompass_Preferred
PKD1/PKC μ Colorimetric Cell-Based ELISA Kit for detecting PKD1/PKC μ in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15139. Gene ID: 5587. Target Synonyms: KPCD1; PKC-mu; PKCM; PKD; PRKCM; PRKD1; Protein kinase C; mu type; Protein kinase D; kinase PKD1; nPKC-mu. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0742