Learn More
Biomatik Corporation PSEN1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50508
Encompass_Preferred
PSEN1 Colorimetric Cell-Based ELISA Kit for detecting PSEN1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49768. Gene ID: 5663. Target Synonyms: AD3; AD3H; presenilin 1; PS-1; PS1; PSN1; PSNL1; S182 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0756