Learn More
Biomatik Corporation S1P Receptor EDG1 Colorimetric Cell-Based ELISA Kit
Catalog No. 502290775
Encompass_Preferred
S1P Receptor EDG1 Colorimetric Cell-Based ELISA Kit for detecting S1P Receptor EDG1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21453. Gene ID: 1901. Target Synonyms: Sphingosine 1-phosphate receptor Edg-1; Sphingosine 1-phosphate receptor 1; S1P1; EDG1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-0775 Supplier Biomatik Corporation Supplier No. EKA50530
S1P Receptor EDG1 Colorimetric Cell-Based ELISA Kit for detecting S1P Receptor EDG1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21453. Gene ID: 1901. Target Synonyms: Sphingosine 1-phosphate receptor Edg-1; Sphingosine 1-phosphate receptor 1; S1P1; EDG1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.