Learn More
Biomatik Corporation SENP2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50533
Encompass_Preferred
SENP2 Colorimetric Cell-Based ELISA Kit for detecting SENP2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9HC62. Gene ID: 59343. Target Synonyms: Sentrin-specific protease 2; Sentrin/SUMO-specific protease SENP2; SMT3-specific isopeptidase 2; Axam2; KIAA1331. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0778